Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01810842_g1
          See other CADN GE Assays ›
          Gene Symbol:
          CadN
          Gene Name:
          Cadherin-N
          Gene Aliases:
          B, CG7100, CT21941, Cad-N, D-cad, DN, DN-CAD, DN-CAd, DN-Cad, DN-cad, DNCad, DNcad, Dmel\CG7100, Dmel_CG7100, N-Cad, N-Cadh, N-cad, N-cadherin, NCAD, NCad, Ncad, S(DmcycE[JP])2.12, anon-EST:CL32, cad1, cadN, l(2)36Da, l(2)Bld, n-cad, ncad
          Chromosome Location:
          Chr.2L: 17647053 - 17778340 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0015609
          Assay ID Dm01810842_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CadN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001032106.2 NP_001027277.1 5 - 6 3615 71
          NM_001032107.3 NP_001027278.1 5 - 6 3615 71
          NM_001032108.2 NP_001027279.1 5 - 6 3615 71
          NM_001032109.3 NP_001027280.1 5 - 6 3615 71
          NM_165224.2 NP_724068.1 5 - 6 3615 71
          NM_165225.2 NP_724069.1 5 - 6 3615 71
          NM_165226.2 NP_724070.1 5 - 6 3615 71
          NM_165227.3 NP_724071.1 7 - 8 3771 71
          NM_165228.2 NP_724072.1 5 - 6 3615 71
          NM_165229.2 NP_724073.1 5 - 6 3615 71
          NM_165230.2 NP_724074.1 5 - 6 3615 71
          NM_165231.3 NP_724075.1 5 - 6 3615 71

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          axon guidance
          axon target recognition
          axonal fasciculation
          ommatidial rotation
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          retinal ganglion cell axon guidance
          cell-cell adhesion mediated by cadherin
          R8 cell development
          R7 cell development
          axon extension
          regulation of dendrite morphogenesis
          regulation of axon extension involved in axon guidance
          axon extension involved in axon guidance
          negative regulation of dendrite morphogenesis
          cell-cell adhesion
          calcium ion binding
          protein binding
          protein homodimerization activity
          cadherin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline