Search
Search

didum| Gene Symbol: didum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057838.4 | NP_477186.1 | 4 - 5 | 1902 | 150 | |
| NM_165535.2 | NP_724569.1 | 4 - 5 | 1902 | 150 | ||
| NM_165536.3 | NP_724570.1 | 4 - 5 | 1902 | 150 | ||
Set Membership: |
|