Search
Search

rgr| Gene Symbol: rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169618.2 | NP_001163089.2 | 5 - 6 | 1865 | 72 | |
| NM_165611.3 | NP_724690.1 | 4 - 5 | 1766 | 72 | ||
| GenBank mRNA | AF353512.1 | - | 4 - 5 | 1714 | 72 | |
| BT150319.1 | - | 4 - 5 | 1737 | 72 | ||
Set Membership: |
|