Search
Search

tea| Gene Symbol: tea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169634.2 | NP_001163105.1 | 3 - 4 | 844 | 95 | |
| NM_001169635.2 | NP_001163106.1 | 3 - 4 | 844 | 95 | ||
| NM_136689.3 | NP_610533.1 | 3 - 4 | 844 | 95 | ||
| NM_165719.3 | NP_724855.2 | 3 - 4 | 844 | 95 | ||
| GenBank mRNA | BT133093.1 | - | 3 - 4 | 799 | 95 | |
Set Membership: |
|