Search
Search

Pfk| Gene Symbol: Pfk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078952.3 | NP_523676.1 | 8 - 9 | 1341 | 85 | |
| NM_165746.3 | NP_724890.1 | 4 - 5 | 1312 | 85 | ||
| GenBank mRNA | AF145673.1 | - | 8 - 9 | 1341 | 85 | |
| BT133189.1 | - | 4 - 5 | 905 | 85 | ||
Set Membership: |
|