Search
Search

Top3beta| Gene Symbol: Top3beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298009.1 | NP_001284938.1 | 3 - 4 | 2156 | 73 | |
| NM_078504.4 | NP_511059.2 | 3 - 4 | 2156 | 73 | ||
| GenBank mRNA | BT016116.1 | - | 3 - 4 | 2118 | 73 | |
Set Membership: |
|