Search
Search

nod| Gene Symbol: nod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298200.1 | NP_001285129.1 | 3 - 4 | 1004 | 67 | |
| NM_078570.3 | NP_511125.2 | 3 - 4 | 1004 | 67 | ||
| GenBank mRNA | AY050238.1 | - | 3 - 4 | 958 | 67 | |
Set Membership: |
|