Search
Search

LIMK1| Gene Symbol: LIMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169262.1 | NP_001162733.1 | 4 - 5 | 3696 | 86 | |
| NM_001169263.1 | NP_001162734.1 | 3 - 4 | 2882 | 86 | ||
| NM_078584.3 | NP_511139.2 | 5 - 6 | 3762 | 86 | ||
| NM_167326.2 | NP_727621.1 | 4 - 5 | 3659 | 86 | ||
| NM_167327.2 | NP_727622.1 | 3 - 4 | 2949 | 86 | ||
| GenBank mRNA | AY119587.1 | - | 4 - 5 | 3548 | 86 | |
| BT133248.1 | - | 3 - 4 | 2906 | 86 | ||
Set Membership: |
|