Search
Search

NFAT| Gene Symbol: NFAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103504.2 | NP_001096974.1 | 6 - 7 | 2366 | 89 | |
| NM_001169274.2 | NP_001162745.1 | 5 - 6 | 1739 | 89 | ||
| NM_001169275.2 | NP_001162746.1 | 5 - 6 | 1958 | 89 | ||
| NM_001169276.3 | NP_001162747.1 | 5 - 6 | 1739 | 89 | ||
| NM_001272597.2 | NP_001259526.2 | 5 - 6 | 1958 | 89 | ||
| NM_001298290.1 | NP_001285219.1 | 6 - 7 | 1764 | 89 | ||
| NM_078592.3 | NP_511147.2 | 6 - 7 | 2585 | 89 | ||
| GenBank mRNA | AF195496.1 | - | 5 - 6 | 1592 | 89 | |
| BT015267.1 | - | 5 - 6 | 1592 | 89 | ||
| BT132967.1 | - | 5 - 6 | 1731 | 89 | ||
Set Membership: |
|