Search
Search

mRpS25| Gene Symbol: mRpS25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078598.2 | NP_511153.1 | 1 - 2 | 222 | 83 | |
| GenBank mRNA | BT024442.1 | - | 1 - 2 | 167 | 83 | |
| BT029396.1 | - | 1 - 2 | 222 | 83 | ||
Set Membership: |
|