Search
Search

Cngl| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258761.3 | NP_001245690.1 | 14 - 15 | 4871 | 80 | |
| NM_001258762.3 | NP_001245691.1 | 15 - 16 | 4916 | 80 | ||
| NM_001272642.2 | NP_001259571.2 | 15 - 16 | 4305 | 80 | ||
| NM_001370020.1 | NP_001356908.1 | 15 - 16 | 4305 | 80 | ||
| NM_167441.5 | NP_727848.3 | 16 - 17 | 4916 | 80 | ||
Set Membership: |
|