Search
Search

Cngl| Gene Symbol: Cngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258761.3 | NP_001245690.1 | 5 - 6 | 3014 | 65 | |
| NM_001258762.3 | NP_001245691.1 | 6 - 7 | 3059 | 65 | ||
| NM_001272642.2 | NP_001259571.2 | 6 - 7 | 2532 | 65 | ||
| NM_001370020.1 | NP_001356908.1 | 6 - 7 | 2532 | 65 | ||
| NM_167441.5 | NP_727848.3 | 6 - 7 | 3059 | 65 | ||
Set Membership: |
|