Search
Search

sd| Gene Symbol: sd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103520.3 | NP_001096990.1 | 2 - 3 | 764 | 61 | |
| NM_001103521.3 | NP_001096991.1 | 2 - 3 | 559 | 61 | ||
| NM_001103522.3 | NP_001096992.1 | 2 - 3 | 651 | 61 | ||
| NM_001169290.2 | NP_001162761.1 | 2 - 3 | 764 | 61 | ||
| NM_001169291.2 | NP_001162762.1 | 2 - 3 | 807 | 61 | ||
| NM_001258768.1 | NP_001245697.1 | 2 - 3 | 625 | 61 | ||
| NM_001258770.3 | NP_001245699.1 | 2 - 3 | 899 | 61 | ||
| NM_001272653.2 | NP_001259582.1 | 3 - 4 | 815 | 61 | ||
| NM_001272654.2 | NP_001259583.1 | 2 - 3 | 903 | 61 | ||
| NM_001382196.1 | NP_001368984.1 | 2 - 3 | 899 | 61 | ||
| NM_078614.5 | NP_511169.1 | 3 - 4 | 706 | 61 | ||
| GenBank mRNA | BT133072.1 | - | 2 - 3 | 761 | 61 | |
| M83787.1 | - | 3 - 4 | 704 | 61 | ||
Set Membership: |
|