Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01813726_m1
          See other PARA GE Assays ›
          Gene Symbol:
          para
          Gene Name:
          paralytic
          Gene Aliases:
          CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
          Chromosome Location:
          Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0285944
          Assay ID Dm01813726_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: para
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042815.2 NP_001036280.1 18 - 19 3012 95
          NM_001201671.1 NP_001188600.1 18 - 19 3012 95
          NM_001201673.1 NP_001188602.1 17 - 18 2949 95
          NM_001201674.1 NP_001188603.1 17 - 18 2979 95
          NM_001201675.1 NP_001188604.1 17 - 18 2925 95
          NM_001201679.1 NP_001188608.1 18 - 19 2988 95
          NM_001201680.1 NP_001188609.1 17 - 18 2949 95
          NM_001201681.1 NP_001188610.1 17 - 18 2925 95
          NM_001201682.1 NP_001188611.1 18 - 19 3012 95
          NM_001201683.1 NP_001188612.1 18 - 19 2916 95
          NM_001201686.1 NP_001188615.1 18 - 19 2988 95
          NM_001201687.1 NP_001188616.1 18 - 19 3012 95
          NM_001201688.1 NP_001188617.1 17 - 18 2949 95
          NM_001201689.1 NP_001188618.1 17 - 18 2925 95
          NM_001201693.1 NP_001188622.1 18 - 19 2940 95
          NM_001201695.2 NP_001188624.1 18 - 19 2940 95
          NM_001201696.1 NP_001188625.1 16 - 17 2916 95
          NM_001201697.1 NP_001188626.1 18 - 19 3012 95
          NM_001201698.1 NP_001188627.1 17 - 18 2979 95
          NM_001201699.1 NP_001188628.1 16 - 17 2916 95
          NM_001201700.1 NP_001188629.1 17 - 18 2979 95
          NM_001201701.1 NP_001188630.1 16 - 17 2916 95
          NM_001201702.1 NP_001188631.1 17 - 18 2979 95
          NM_001201703.3 NP_001188632.1 18 - 19 3502 95
          NM_001201704.1 NP_001188633.1 16 - 17 2916 95
          NM_001201705.1 NP_001188634.1 17 - 18 2907 95
          NM_001201706.1 NP_001188635.1 18 - 19 3012 95
          NM_001201707.1 NP_001188636.1 18 - 19 2988 95
          NM_001201708.1 NP_001188637.1 17 - 18 2949 95
          NM_001201709.1 NP_001188638.1 17 - 18 2925 95
          NM_001201710.1 NP_001188639.1 14 - 15 2695 95
          NM_001201711.1 NP_001188640.1 16 - 17 2850 95
          NM_001201712.1 NP_001188641.1 15 - 16 2787 95
          NM_001201713.1 NP_001188642.1 15 - 16 2787 95
          NM_001201714.1 NP_001188643.1 15 - 16 2763 95
          NM_001201715.1 NP_001188644.1 16 - 17 2844 95
          NM_001201716.1 NP_001188645.1 17 - 18 2955 95
          NM_001201717.1 NP_001188646.1 17 - 18 2979 95
          NM_001201718.1 NP_001188647.1 16 - 17 2916 95
          NM_001201720.1 NP_001188649.1 17 - 18 2979 95
          NM_001201722.1 NP_001188651.1 17 - 18 2979 95
          NM_001272690.1 NP_001259619.1 18 - 19 3012 95
          NM_001298403.1 NP_001285332.1 18 - 19 3165 95
          NM_001298404.1 NP_001285333.1 17 - 18 2976 95
          NM_078647.4 NP_523371.2 18 - 19 3012 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          mechanosensory behavior
          response to mechanical stimulus
          neuronal action potential
          sodium ion transmembrane transport
          male courtship behavior, veined wing generated song production
          regulation of postsynaptic membrane potential
          membrane depolarization during action potential
          voltage-gated sodium channel activity
          cation channel activity
          sodium channel activity
          calcium ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline