Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01813747_g1
See other PARA GE Assays ›
Gene Symbol:
para
Gene Name:
paralytic
Gene Aliases:
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location:
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0285944
Assay ID
Dm01813747_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: para
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042815.2
NP_001036280.1
9 - 10
1314
75
NM_001042816.2
NP_001036281.1
9 - 10
1314
75
NM_001144733.2
NP_001138205.1
8 - 9
1281
75
NM_001201671.1
NP_001188600.1
9 - 10
1314
75
NM_001201672.1
NP_001188601.1
9 - 10
1314
75
NM_001201673.1
NP_001188602.1
9 - 10
1314
75
NM_001201674.1
NP_001188603.1
8 - 9
1281
75
NM_001201675.1
NP_001188604.1
9 - 10
1314
75
NM_001201676.1
NP_001188605.1
9 - 10
1314
75
NM_001201677.1
NP_001188606.1
9 - 10
1314
75
NM_001201678.1
NP_001188607.1
9 - 10
1314
75
NM_001201679.1
NP_001188608.1
9 - 10
1314
75
NM_001201680.1
NP_001188609.1
9 - 10
1314
75
NM_001201681.1
NP_001188610.1
9 - 10
1314
75
NM_001201682.1
NP_001188611.1
9 - 10
1314
75
NM_001201683.1
NP_001188612.1
9 - 10
1314
75
NM_001201684.1
NP_001188613.1
9 - 10
1314
75
NM_001201685.1
NP_001188614.1
9 - 10
1314
75
NM_001201686.1
NP_001188615.1
9 - 10
1314
75
NM_001201687.1
NP_001188616.1
9 - 10
1314
75
NM_001201688.1
NP_001188617.1
9 - 10
1314
75
NM_001201689.1
NP_001188618.1
9 - 10
1314
75
NM_001201690.1
NP_001188619.1
9 - 10
1314
75
NM_001201691.1
NP_001188620.1
9 - 10
1314
75
NM_001201692.1
NP_001188621.1
9 - 10
1314
75
NM_001201693.1
NP_001188622.1
9 - 10
1314
75
NM_001201694.1
NP_001188623.1
9 - 10
1314
75
NM_001201695.2
NP_001188624.1
9 - 10
1314
75
NM_001201696.1
NP_001188625.1
8 - 9
1281
75
NM_001201697.1
NP_001188626.1
9 - 10
1314
75
NM_001201698.1
NP_001188627.1
8 - 9
1281
75
NM_001201699.1
NP_001188628.1
8 - 9
1281
75
NM_001201700.1
NP_001188629.1
8 - 9
1281
75
NM_001201701.1
NP_001188630.1
8 - 9
1281
75
NM_001201702.1
NP_001188631.1
8 - 9
1281
75
NM_001201703.3
NP_001188632.1
9 - 10
1804
75
NM_001201704.1
NP_001188633.1
8 - 9
1281
75
NM_001201705.1
NP_001188634.1
8 - 9
1281
75
NM_001201706.1
NP_001188635.1
9 - 10
1314
75
NM_001201707.1
NP_001188636.1
9 - 10
1314
75
NM_001201708.1
NP_001188637.1
9 - 10
1314
75
NM_001201709.1
NP_001188638.1
9 - 10
1314
75
NM_001201710.1
NP_001188639.1
6 - 7
1060
75
NM_001201711.1
NP_001188640.1
7 - 8
1152
75
NM_001201712.1
NP_001188641.1
7 - 8
1152
75
NM_001201713.1
NP_001188642.1
7 - 8
1152
75
NM_001201714.1
NP_001188643.1
7 - 8
1152
75
NM_001201715.1
NP_001188644.1
8 - 9
1281
75
NM_001201716.1
NP_001188645.1
8 - 9
1281
75
NM_001201717.1
NP_001188646.1
8 - 9
1281
75
NM_001201718.1
NP_001188647.1
8 - 9
1281
75
NM_001201719.1
NP_001188648.1
8 - 9
1281
75
NM_001201720.1
NP_001188649.1
8 - 9
1281
75
NM_001201721.1
NP_001188650.1
8 - 9
1281
75
NM_001201722.1
NP_001188651.1
8 - 9
1281
75
NM_001201723.1
NP_001188652.1
8 - 9
1281
75
NM_001272690.1
NP_001259619.1
9 - 10
1314
75
NM_001298403.1
NP_001285332.1
9 - 10
1467
75
NM_001298404.1
NP_001285333.1
8 - 9
1278
75
NM_078647.4
NP_523371.2
9 - 10
1314
75
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Drosophila melanogaster
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
mechanosensory behavior
response to mechanical stimulus
neuronal action potential
sodium ion transmembrane transport
male courtship behavior, veined wing generated song production
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
calcium ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline