Search
Search

mbt| Gene Symbol: mbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078651.4 | NP_523375.2 | 2 - 3 | 1951 | 87 | |
| GenBank mRNA | AJ011578.1 | - | 2 - 3 | 1900 | 87 | |
| BT044204.1 | - | 2 - 3 | 1924 | 87 | ||
Set Membership: |
|