Search
Search

baz| Gene Symbol: baz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042818.4 | NP_001036283.1 | 5 - 6 | 7459 | 59 | |
| NM_001298435.2 | NP_001285364.1 | 3 - 4 | 3505 | 59 | ||
| NM_001298436.2 | NP_001285365.1 | 5 - 6 | 7459 | 59 | ||
| NM_001347807.1 | NP_001334669.1 | 3 - 4 | 3505 | 59 | ||
| GenBank mRNA | BT032834.1 | - | 3 - 4 | 2576 | 59 | |
Set Membership: |
|