Search
Search

beta-Spec| Gene Symbol: beta-Spec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272731.1 | NP_001259660.1 | 6 - 7 | 6933 | 109 | |
| NM_001272732.1 | NP_001259661.1 | 6 - 7 | 7111 | 109 | ||
| NM_078664.3 | NP_523388.1 | 5 - 6 | 6886 | 109 | ||
Set Membership: |
|