Search
Search

pcm| Gene Symbol: pcm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169325.2 | NP_001162796.1 | 2 - 3 | 1372 | 117 | |
| NM_078684.4 | NP_523408.2 | 2 - 3 | 1372 | 117 | ||
| GenBank mRNA | AY051675.1 | - | 2 - 3 | 1312 | 117 | |
Set Membership: |
|