Search
Search

Grip84| Gene Symbol: Grip84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169326.2 | NP_001162797.1 | 2 - 3 | 888 | 61 | |
| NM_001169327.2 | NP_001162798.1 | 2 - 3 | 888 | 61 | ||
| NM_001298515.1 | NP_001285444.1 | 1 - 2 | 828 | 61 | ||
| NM_078685.3 | NP_523409.1 | 1 - 2 | 828 | 61 | ||
| NM_167662.2 | NP_728264.1 | 2 - 3 | 888 | 61 | ||
| NM_167663.2 | NP_728265.1 | 1 - 2 | 828 | 61 | ||
| GenBank mRNA | AF118379.1 | - | 1 - 2 | 652 | 61 | |
| BT150451.1 | - | 2 - 3 | 721 | 61 | ||
Set Membership: |
|