Search
Search

Zw| Gene Symbol: Zw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298527.1 | NP_001285456.1 | 2 - 3 | 337 | 77 | |
| NM_078687.2 | NP_523411.1 | 2 - 3 | 632 | 77 | ||
| NM_167676.2 | NP_728287.1 | 2 - 3 | 609 | 77 | ||
| GenBank mRNA | BT053690.1 | - | 2 - 3 | 584 | 77 | |
Set Membership: |
|