Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01814108_g1
          See other L(2)GL GE Assays ›
          Gene Symbol:
          l(2)gl
          Gene Name:
          lethal (2) giant larvae
          Gene Aliases:
          CG2671, D-LGL, Dmel\CG2671, Dmel_CG2671, L(2)GL, L(2)gl, LGL, Lgl, MENE (2L)-B, dlgl, gl, l(2), l(2) giant larva, l(2)giant larvae, l-gl, l[[2]]gl, lgl, p127, p127[l(2)gl], p127l(2)gl
          Chromosome Location:
          Chr.2L: 9839 - 21376 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0002121
          Assay ID Dm01814108_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: l(2)gl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001258872.2 NP_001245801.1 4 - 5 1323 85
          NM_001258873.2 NP_001245802.1 3 - 4 1187 85
          NM_001258874.2 NP_001245803.1 4 - 5 1398 85
          NM_001258875.2 NP_001245804.1 5 - 6 1536 85
          NM_001272858.1 NP_001259787.1 4 - 5 1398 85
          NM_078715.4 NP_523439.2 3 - 4 1256 85
          NM_164348.3 NP_722583.1 3 - 4 1163 85
          NM_164349.3 NP_722584.1 4 - 5 1399 85
          NM_164350.3 NP_722585.1 3 - 4 1238 85
          NM_164351.3 NP_722586.1 3 - 4 1233 85
          NM_164352.3 NP_722587.1 4 - 5 1090 85
          GenBank mRNA AY051654.1 - 3 - 4 1246 85
          BT100129.1 - 3 - 4 1159 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          negative regulation of protein kinase activity
          oocyte anterior/posterior axis specification
          dorsal closure
          nervous system development
          negative regulation of neuroblast proliferation
          sensory organ development
          regulation of Notch signaling pathway
          oocyte microtubule cytoskeleton organization
          morphogenesis of follicular epithelium
          establishment or maintenance of polarity of follicular epithelium
          sensory organ precursor cell fate determination
          septate junction assembly
          actin cytoskeleton organization
          cortical actin cytoskeleton organization
          regulation of establishment or maintenance of cell polarity
          salivary gland histolysis
          ecdysone-mediated induction of salivary gland cell autophagic cell death
          cell competition in a multicellular organism
          chitin-based larval cuticle pattern formation
          positive regulation of hippo signaling
          asymmetric protein localization involved in cell fate determination
          basal protein localization
          apical protein localization
          establishment of protein localization
          establishment or maintenance of neuroblast polarity
          R8 cell fate specification
          negative regulation of imaginal disc growth
          negative regulation of Notch signaling pathway
          epidermis morphogenesis
          regulation of protein secretion
          establishment of spindle orientation
          localization within membrane
          asymmetric neuroblast division
          protein localization to cell cortex
          establishment of epithelial cell planar polarity
          symmetric cell division
          regulation of intracellular mRNA localization
          SNARE binding
          protein kinase inhibitor activity
          GTPase activator activity
          myosin binding
          Rab GTPase binding
          protein kinase binding
          syntaxin binding
          myosin II binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline