Search
Search

Rab5| Gene Symbol: Rab5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272996.1 | NP_001259925.1 | 3 - 4 | 679 | 57 | |
| NM_001272997.1 | NP_001259926.1 | 3 - 4 | 670 | 57 | ||
| NM_001272998.1 | NP_001259927.1 | 3 - 4 | 669 | 57 | ||
| NM_078733.4 | NP_523457.1 | 3 - 4 | 652 | 57 | ||
| NM_164475.2 | NP_722795.1 | 3 - 4 | 659 | 57 | ||
| NM_164476.2 | NP_722796.1 | 3 - 4 | 765 | 57 | ||
| NM_164477.3 | NP_722797.1 | 3 - 4 | 961 | 57 | ||
| NM_164478.2 | NP_722798.1 | 3 - 4 | 633 | 57 | ||
| NM_164479.2 | NP_722799.1 | 3 - 4 | 617 | 57 | ||
| GenBank mRNA | AB035353.1 | - | 3 - 4 | 667 | 57 | |
| AB035671.1 | - | 3 - 4 | 638 | 57 | ||
| AY060343.1 | - | 3 - 4 | 937 | 57 | ||
| AY081180.1 | - | 2 - 3 | 537 | 57 | ||
| AY081181.1 | - | 3 - 4 | 621 | 57 | ||
Set Membership: |
|