Search
Search

lilli| Gene Symbol: lilli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144300.2 | NP_001137772.1 | 5 - 6 | 4205 | 73 | |
| NM_001144301.1 | NP_001137773.1 | 6 - 7 | 4545 | 73 | ||
| NM_001144302.2 | NP_001137774.1 | 5 - 6 | 4314 | 73 | ||
| NM_001144303.1 | NP_001137775.1 | 7 - 8 | 5849 | 73 | ||
| NM_001273044.1 | NP_001259973.1 | 6 - 7 | 6002 | 73 | ||
| NM_001347818.1 | NP_001334729.1 | 6 - 7 | 6002 | 73 | ||
| NM_078740.3 | NP_523464.1 | 7 - 8 | 6765 | 73 | ||
| NM_164516.2 | NP_722863.1 | 8 - 9 | 6933 | 73 | ||
| NM_164517.1 | NP_722864.1 | 7 - 8 | 5654 | 73 | ||
| GenBank mRNA | AF289034.1 | - | 8 - 9 | 6910 | 73 | |
| AF293971.1 | - | 7 - 8 | 6739 | 73 | ||
Set Membership: |
|