Search
Search

wg| Gene Symbol: wg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078778.5 | NP_523502.1 | 1 - 2 | 546 | 75 | |
| GenBank mRNA | BT133499.1 | - | 1 - 2 | 544 | 75 | |
| M17230.1 | - | 1 - 2 | 530 | 75 | ||
Set Membership: |
|