Search
Search

sick| Gene Symbol: sick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103713.4 | NP_001097183.1 | 11 - 12 | 5501 | 66 | |
| NM_001273674.1 | NP_001260603.1 | 13 - 14 | 6364 | 66 | ||
| NM_001273675.1 | NP_001260604.1 | 3 - 4 | 3657 | 66 | ||
| NM_001273676.1 | NP_001260605.1 | 11 - 12 | 5617 | 66 | ||
| NM_001273677.1 | NP_001260606.1 | 12 - 13 | 7383 | 66 | ||
| NM_001273678.1 | NP_001260607.1 | 12 - 13 | 8069 | 66 | ||
| NM_001273679.1 | NP_001260608.1 | 8 - 9 | 3834 | 66 | ||
| NM_001273680.2 | NP_001260609.1 | 14 - 15 | 6296 | 66 | ||
| NM_001273681.1 | NP_001260610.1 | 11 - 12 | 5822 | 66 | ||
| NM_136167.3 | NP_610011.2 | 3 - 4 | 3556 | 66 | ||
| NM_206007.5 | NP_995729.2 | 13 - 14 | 6269 | 66 | ||
| NM_206008.4 | NP_995730.2 | 13 - 14 | 8236 | 66 | ||
Set Membership: |
|