Search
Search

grh| Gene Symbol: grh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259472.1 | NP_001246401.1 | 2 - 3 | 385 | 118 | |
| NM_057496.5 | NP_476844.2 | 4 - 5 | 2691 | 118 | ||
| NM_057497.4 | NP_476845.2 | 4 - 5 | 3312 | 118 | ||
| NM_166252.3 | NP_725725.1 | 2 - 3 | 379 | 118 | ||
Set Membership: |
|