Search
Search

SdhA| Gene Symbol: SdhA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057862.5 | NP_477210.1 | 2 - 3 | 1877 | 71 | |
| NM_166343.3 | NP_725881.1 | 3 - 4 | 1744 | 71 | ||
| NM_166344.3 | NP_725882.1 | 3 - 4 | 1790 | 71 | ||
| GenBank mRNA | AY051472.1 | - | 2 - 3 | 1842 | 71 | |
| BT099627.1 | - | 3 - 4 | 1830 | 71 | ||
Set Membership: |
|