Search
Search

hrg| Gene Symbol: hrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043097.3 | NP_001036562.1 | 3 - 4 | 1473 | 73 | |
| NM_001274168.2 | NP_001261097.1 | 3 - 4 | 1602 | 73 | ||
| NM_080529.4 | NP_536790.1 | 3 - 4 | 1473 | 73 | ||
| NM_166372.3 | NP_725919.1 | 3 - 4 | 1166 | 73 | ||
| GenBank mRNA | AB018431.1 | - | 3 - 4 | 1462 | 73 | |
| AF231704.1 | - | 4 - 5 | 1432 | 73 | ||
| BT001541.1 | - | 3 - 4 | 1159 | 73 | ||
| BT050444.1 | - | 3 - 4 | 1470 | 73 | ||
Set Membership: |
|