Search
Search

CG11200| Gene Symbol: CG11200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299701.1 | NP_001286630.1 | 3 - 4 | 438 | 60 | |
| NM_137627.4 | NP_611471.1 | 4 - 5 | 791 | 60 | ||
| NM_166388.3 | NP_725952.1 | 3 - 4 | 550 | 60 | ||
| GenBank mRNA | AY070514.1 | - | 4 - 5 | 779 | 60 | |
Set Membership: |
|