Search
Search

GABA-B-R1| Gene Symbol: GABA-B-R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259104.2 | NP_001246033.1 | 6 - 7 | 2009 | 89 | |
| NM_078845.5 | NP_523569.3 | 6 - 7 | 2009 | 89 | ||
| GenBank mRNA | BT031099.1 | - | 6 - 7 | 1205 | 89 | |
Set Membership: |
|