Search
Search

Tsp42Ed| Gene Symbol: Tsp42Ed | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299223.1 | NP_001286152.1 | 3 - 4 | 500 | 66 | |
| NM_078906.3 | NP_523630.1 | 3 - 4 | 500 | 66 | ||
| GenBank mRNA | AF220042.1 | - | 3 - 4 | 486 | 66 | |
| AY102700.1 | - | 3 - 4 | 614 | 66 | ||
Set Membership: |
|