Search
Search

pds5| Gene Symbol: pds5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259342.2 | NP_001246271.1 | 6 - 7 | 2114 | 69 | |
| NM_136875.4 | NP_610719.1 | 6 - 7 | 1856 | 69 | ||
| GenBank mRNA | BT053744.1 | - | 6 - 7 | 1836 | 69 | |
Set Membership: |
|