Search
Search

eEF5| Gene Symbol: eEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138034.3 | NP_611878.1 | 3 - 4 | 467 | 81 | |
| NM_166654.2 | NP_726411.1 | 3 - 4 | 518 | 81 | ||
| GenBank mRNA | AY071396.1 | - | 3 - 4 | 496 | 81 | |
| KX531679.1 | - | 2 - 3 | 411 | 81 | ||
Set Membership: |
|