Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01820463_m1
          See other ZIP GE Assays ›
          Gene Symbol:
          zip
          Gene Name:
          zipper
          Gene Aliases:
          CG15792, DROMHC, Dm nmII, Dmel\CG15792, Dmel_CG15792, DmnmII, Dronm-MII, E(br), MHC, Mhc-c, Mhc-c[1], Myo, Myo II, Myo-II, Myo-II HC, Myo15, Myo2, MyoII, NMM, NMMII, ZIP, Zip, anon-WO0140519.37, l(2)02957, l(2)17F1, myo II, myoII, myosin-II, nmMHC, nmy-2, zip/MyoII, zpr
          Chromosome Location:
          Chr.2R: 24990570 - 25011965 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0265434
          Assay ID Dm01820463_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: zip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014552.3 NP_001014552.1 3 - 4 600 92
          NM_001014553.3 NP_001014553.1 3 - 4 536 92
          NM_001202079.1 NP_001189008.1 3 - 4 563 92
          NM_001202080.2 NP_001189009.1 3 - 4 536 92
          NM_001202081.1 NP_001189010.1 3 - 4 557 92
          NM_001202082.2 NP_001189011.1 3 - 4 536 92
          NM_001259587.2 NP_001246516.1 3 - 4 600 92
          NM_079136.4 NP_523860.2 3 - 4 600 92
          NM_166704.4 NP_726506.1 3 - 4 536 92

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Drosophila melanogaster

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          muscle contraction
          border follicle cell migration
          dorsal closure
          dorsal closure, spreading of leading edge cells
          salivary gland morphogenesis
          Malpighian tubule morphogenesis
          eye-antennal disc morphogenesis
          anterior midgut development
          head involution
          muscle attachment
          myofibril assembly
          myosin II filament assembly
          maintenance of protein location in cell
          cuticle pattern formation
          regulation of tube length, open tracheal system
          spiracle morphogenesis, open tracheal system
          imaginal disc-derived wing hair organization
          wound healing
          sarcomere organization
          dorsal closure, leading edge cell differentiation
          dorsal closure, amnioserosa morphology change
          compartment boundary maintenance
          morphogenesis of an epithelial fold
          left/right axis specification
          cellular response to mechanical stimulus
          regulation of establishment of planar polarity
          regulation of myoblast fusion
          positive regulation of border follicle cell migration
          motor activity
          protein binding
          ATP binding
          myosin light chain binding
          actin filament binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline