Search
Search

zip| Gene Symbol: zip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014552.3 | NP_001014552.1 | 4 - 5 | 757 | 79 | |
| NM_001014553.3 | NP_001014553.1 | 4 - 5 | 693 | 79 | ||
| NM_001202079.1 | NP_001189008.1 | 4 - 5 | 720 | 79 | ||
| NM_001202080.2 | NP_001189009.1 | 4 - 5 | 693 | 79 | ||
| NM_001202081.1 | NP_001189010.1 | 4 - 5 | 714 | 79 | ||
| NM_001202082.2 | NP_001189011.1 | 4 - 5 | 693 | 79 | ||
| NM_001259587.2 | NP_001246516.1 | 4 - 5 | 757 | 79 | ||
| NM_079136.4 | NP_523860.2 | 4 - 5 | 757 | 79 | ||
| NM_166704.4 | NP_726506.1 | 4 - 5 | 693 | 79 | ||
Set Membership: |
|