Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01820467_g1
See other ZIP GE Assays ›
Gene Symbol:
zip
Gene Name:
zipper
Gene Aliases:
CG15792, DROMHC, Dm nmII, Dmel\CG15792, Dmel_CG15792, DmnmII, Dronm-MII, E(br), MHC, Mhc-c, Mhc-c[1], Myo, Myo II, Myo-II, Myo-II HC, Myo15, Myo2, MyoII, NMM, NMMII, ZIP, Zip, anon-WO0140519.37, l(2)02957, l(2)17F1, myo II, myoII, myosin-II, nmMHC, nmy-2, zip/MyoII, zpr
Chromosome Location:
Chr.2R: 24990570 - 25011965 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0265434
Assay ID
Dm01820467_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: zip
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_079136.4
NP_523860.2
7 - 8
1033
86
NM_166704.4
NP_726506.1
7 - 8
969
86
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Drosophila melanogaster
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
muscle contraction
border follicle cell migration
dorsal closure
dorsal closure, spreading of leading edge cells
salivary gland morphogenesis
Malpighian tubule morphogenesis
eye-antennal disc morphogenesis
anterior midgut development
head involution
muscle attachment
myofibril assembly
myosin II filament assembly
maintenance of protein location in cell
cuticle pattern formation
regulation of tube length, open tracheal system
spiracle morphogenesis, open tracheal system
imaginal disc-derived wing hair organization
wound healing
sarcomere organization
dorsal closure, leading edge cell differentiation
dorsal closure, amnioserosa morphology change
compartment boundary maintenance
morphogenesis of an epithelial fold
left/right axis specification
cellular response to mechanical stimulus
regulation of establishment of planar polarity
regulation of myoblast fusion
positive regulation of border follicle cell migration
motor activity
protein binding
ATP binding
myosin light chain binding
actin filament binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline