Search
Search

gol| Gene Symbol: gol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169829.3 | NP_001163300.1 | 2 - 3 | 1119 | 80 | |
| NM_001259588.2 | NP_001246517.1 | 2 - 3 | 1119 | 80 | ||
| NM_079140.5 | NP_523864.3 | 2 - 3 | 1119 | 80 | ||
| NM_166705.3 | NP_726508.1 | 2 - 3 | 1119 | 80 | ||
| GenBank mRNA | AY069169.1 | - | 2 - 3 | 1092 | 80 | |
Set Membership: |
|