Search
Search

ey| Gene Symbol: ey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014693.2 | NP_001014693.1 | 6 - 7 | 1983 | 75 | |
| NM_001014694.2 | NP_001014694.1 | 5 - 6 | 1423 | 75 | ||
| NM_079889.3 | NP_524628.2 | 5 - 6 | 1803 | 75 | ||
| NM_166789.2 | NP_726607.1 | 5 - 6 | 1375 | 75 | ||
| GenBank mRNA | BT011390.1 | - | 5 - 6 | 1375 | 75 | |
Set Membership: |
|