Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01821914_g1
See other GLUT1 GE Assays ›
Gene Symbol:
Glut1
Gene Name:
Glucose transporter 1
Gene Aliases:
0074/12, CG1086, CG13908, CG43946, DmGLUT1, DmGluT1, Dmel\CG43946, Dmel_CG43946, Dmglut1, GLU, GLUT1, Glt1, Glu T1, Glut-1, glut1, l(3)61Ea, l(3)L1H, l(3)S007412
Chromosome Location:
Chr.3L: 914074 - 993788 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0264574
Assay ID
Dm01821914_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Glut1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001103997.3
NP_001097467.1
6 - 7
1538
63
NM_001103998.3
NP_001097468.1
6 - 7
1538
63
NM_001103999.4
NP_001097469.2
8 - 9
1842
63
NM_001104000.2
NP_001097470.2
6 - 7
1538
63
NM_001274301.1
NP_001261230.1
6 - 7
1538
63
NM_001274302.1
NP_001261231.1
6 - 7
1538
63
NM_001274303.1
NP_001261232.1
9 - 10
1898
63
NM_001274304.1
NP_001261233.1
14 - 15
2629
63
NM_001274305.1
NP_001261234.1
14 - 15
3210
63
NM_001274307.1
NP_001261236.1
11 - 12
2284
63
NM_001274308.1
NP_001261237.1
11 - 12
2284
63
NM_001274309.1
NP_001261238.1
11 - 12
2284
63
NM_001299966.1
NP_001286895.1
6 - 7
1538
63
NM_001299967.1
NP_001286896.1
6 - 7
1538
63
NM_001316474.1
NP_001303403.1
6 - 7
1538
63
NM_079154.3
NP_523878.1
6 - 7
1538
63
NM_138229.2
NP_612073.2
6 - 7
1538
63
NM_167845.3
NP_728557.1
6 - 7
1410
63
NM_167846.3
NP_728558.5
13 - 14
2476
63
GenBank mRNA
AF064703.2
-
6 - 7
1536
63
BT011383.1
-
6 - 7
1506
63
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Drosophila melanogaster
Gene Ontology Categories:
Function(s)
Process(es)
glucose homeostasis
transmembrane transport
positive regulation of peptide hormone secretion
transmembrane transporter activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline