Search
Search

BtbVII| Gene Symbol: BtbVII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144408.3 | NP_001137880.2 | 4 - 5 | 1439 | 60 | |
| NM_001259661.2 | NP_001246590.1 | 4 - 5 | 1439 | 60 | ||
| NM_001259662.1 | NP_001246591.1 | 4 - 5 | 1417 | 60 | ||
| NM_001274426.1 | NP_001261355.1 | 4 - 5 | 1454 | 60 | ||
| NM_079172.4 | NP_523896.2 | 4 - 5 | 1315 | 60 | ||
| NM_139517.3 | NP_647774.2 | 4 - 5 | 1439 | 60 | ||
| NM_167990.3 | NP_728814.1 | 4 - 5 | 1425 | 60 | ||
| GenBank mRNA | AY051895.1 | - | 3 - 4 | 1410 | 60 | |
Set Membership: |
|