Search
Search

Dhc64C| Gene Symbol: Dhc64C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274500.1 | NP_001261429.1 | 11 - 12 | 10198 | 110 | |
| NM_001274501.1 | NP_001261430.1 | 11 - 12 | 10186 | 110 | ||
| NM_001274503.1 | NP_001261432.1 | 12 - 13 | 10225 | 110 | ||
| NM_079205.4 | NP_523929.2 | 10 - 11 | 10159 | 110 | ||
Set Membership: |
|