Search
Search

bol| Gene Symbol: bol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169923.3 | NP_001163394.1 | 6 - 7 | 1173 | 71 | |
| NM_001274685.2 | NP_001261614.1 | 4 - 5 | 1117 | 71 | ||
| NM_001300063.1 | NP_001286992.1 | 4 - 5 | 1536 | 71 | ||
| NM_079265.4 | NP_523989.1 | 4 - 5 | 1117 | 71 | ||
| NM_168318.3 | NP_729456.1 | 5 - 6 | 966 | 71 | ||
| NM_168319.2 | NP_729457.1 | 5 - 6 | 966 | 71 | ||
| NM_168320.2 | NP_729458.1 | 5 - 6 | 1459 | 71 | ||
| GenBank mRNA | BT011495.1 | - | 6 - 7 | 1139 | 71 | |
| U51858.1 | - | 4 - 5 | 1107 | 71 | ||
Set Membership: |
|