Search
Search

I-2| Gene Symbol: I-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104094.2 | NP_001097564.1 | 4 - 5 | 657 | 54 | |
| NM_079289.3 | NP_524013.1 | 4 - 5 | 644 | 54 | ||
| GenBank mRNA | AJ133246.1 | - | 4 - 5 | 591 | 54 | |
| AJ249462.1 | - | 4 - 5 | 625 | 54 | ||
| BT014655.1 | - | 4 - 5 | 606 | 54 | ||
Set Membership: |
|