Search
Search

Adk1| Gene Symbol: Adk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079314.3 | NP_524038.1 | 2 - 3 | 337 | 52 | |
| NM_168493.2 | NP_729792.1 | 3 - 4 | 568 | 52 | ||
| GenBank mRNA | AB050623.1 | - | 1 - 2 | 123 | 52 | |
| AY118287.1 | - | 2 - 3 | 483 | 52 | ||
| BT012445.1 | - | 3 - 4 | 413 | 52 | ||
| BT028794.1 | - | 3 - 4 | 395 | 52 | ||
Set Membership: |
|