Search
Search

eyg| Gene Symbol: eyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014582.2 | NP_001014582.1 | 5 - 6 | 1518 | 63 | |
| NM_001300118.1 | NP_001287047.1 | 4 - 5 | 1585 | 63 | ||
| NM_079318.3 | NP_524042.2 | 4 - 5 | 1585 | 63 | ||
Set Membership: |
|