Search
Search

bru3| Gene Symbol: bru3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202163.2 | NP_001189092.1 | 9 - 10 | 1678 | 63 | |
| NM_001202166.4 | NP_001189095.1 | 9 - 10 | 1702 | 63 | ||
| NM_001202171.2 | NP_001189100.1 | 8 - 9 | 1564 | 63 | ||
| NM_001274878.1 | NP_001261807.1 | 11 - 12 | 2062 | 63 | ||
| NM_001274879.1 | NP_001261808.1 | 11 - 12 | 1798 | 63 | ||
| NM_001382115.1 | NP_001369049.1 | 9 - 10 | 1678 | 63 | ||
| NM_001382116.1 | NP_001369050.1 | 9 - 10 | 1678 | 63 | ||
| NM_001382117.1 | NP_001369051.1 | 10 - 11 | 1663 | 63 | ||
| NM_079335.6 | NP_524059.4 | 8 - 9 | 1543 | 63 | ||
| NM_168557.3 | NP_729923.1 | 10 - 11 | 1720 | 63 | ||
| GenBank mRNA | AY060398.1 | - | 10 - 11 | 1720 | 63 | |
Set Membership: |
|