Search
Search

Eig71Ed| Gene Symbol: Eig71Ed | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079367.4 | NP_524091.1 | 2 - 3 | 295 | 73 | |
| GenBank mRNA | AY119014.1 | - | 2 - 3 | 275 | 73 | |
| KX531994.1 | - | 2 - 3 | 234 | 73 | ||
| U24243.1 | - | 2 - 3 | 288 | 73 | ||
Set Membership: |
|