Search
Search

Mipp1| Gene Symbol: Mipp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300153.1 | NP_001287082.1 | 2 - 3 | 388 | 78 | |
| NM_079385.4 | NP_524109.2 | 2 - 3 | 388 | 78 | ||
| NM_168672.3 | NP_730177.1 | 2 - 3 | 329 | 78 | ||
| GenBank mRNA | AY071426.1 | - | 2 - 3 | 325 | 78 | |
Set Membership: |
|