Search
Search

Dab| Gene Symbol: Dab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169984.3 | NP_001163455.2 | 4 - 5 | 1941 | 61 | |
| NM_001259872.1 | NP_001246801.1 | 4 - 5 | 2089 | 61 | ||
| NM_001275038.1 | NP_001261967.1 | 4 - 5 | 2089 | 61 | ||
| NM_001275040.1 | NP_001261969.1 | 4 - 5 | 1941 | 61 | ||
| NM_079395.4 | NP_524119.2 | 4 - 5 | 1941 | 61 | ||
Set Membership: |
|